2-Phenylethanol ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
P0084-500ML
P0084-25ML
TCP0084-500MLEA
52.8
USD
TCP0084-500ML
TCP0084-025ML
2-Phenylethanol ≥98.0%
2-Phenylethanol
CAS Number: 60-12-8
MDL Number: MFCD00002886
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Melting point (°C): -27
Flash Point (°C): 108
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: Benzyl Carbinol, Phenethyl Alcohol, 2-Phenylethanol
Formula:
C₆H₅CH₂CH₂OH MW: 122.17 g/mol Boiling Pt: 219 °C Melting Pt: –27 °C Density: 1.02 (20 °C) Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002886 CAS Number: 60-12-8 EINECS: 200-456-2 UN: 2810 ADR: 6.1,III Merck Index: 12,07370 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...