[(Oxido)phenyl(trifluoromethyl)-lambda-4-sulfanylidene]dimethylammonium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

O0367-1G O0367-200MG
TCO0367-1GEA 522 USD
TCO0367-1G TCO0367-200MG
[(Oxido)phenyl(trifluoromethyl)-lambda-4-sulfanylidene]dimethylammonium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
[(Oxido)phenyl(trifluoromethyl)-lambda-4-sulfanylidene]dimethylammonium tetrafluoroborate

CAS Number: 1046786-08-6
Molecular Formula: C9H11BF7NOS
Molecular Weight: 325.05
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: Shibata Reagent I

Formula: C₉H₁₁BF₇NOS
CAS Number: 1046786-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f18d6064344c9ff2032dee0abbcd437796c8026f&vid=f18d6064344c9ff2032dee0abbcd437796c8026f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR