Octadecyl-3-mercaptopropionate ≥85.0% (contains ca. 12% Hexadecyl 3-Mercaptopropionate)

Supplier: TCI America

Synonyms: Stearyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Stearyl Ester, 3-Mercaptopropionic Acid Octadecyl Ester

O0362-500G O0362-25G
TCO0362-500GEA 458 USD
TCO0362-500G TCO0362-25G
Octadecyl-3-mercaptopropionate ≥85.0% (contains ca. 12% Hexadecyl 3-Mercaptopropionate)
Octadecyl-3-mercaptopropionate

CAS Number: 31778-15-1
MDL Number: MFCD00046844
Molecular Formula: C21H42O2S
Molecular Weight: 358.63
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Freezing point (°C): 28


Other Product Names / Synonyms: 3-Mercaptopropionic Acid Octadecyl Ester, Stearyl 3-Mercaptopropionate, 3-Mercaptopropionic Acid Stearyl Ester

Formula: C₂₁H₄₂O₂S
MW: 358.63 g/mol
MDL Number: MFCD00046844
CAS Number: 31778-15-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440ff06beaf85cd4fc67d928d2ecc57e85aea954&vid=440ff06beaf85cd4fc67d928d2ecc57e85aea954&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >85.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR