Omeprazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 6-Methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole

O0359-25G O0359-5G
TCO0359-25GEA 356 USD
TCO0359-25G TCO0359-5G
Omeprazole ≥98.0% (by HPLC, titration analysis)
Omeprazole

CAS Number: 73590-58-6 MDL Number: MFCD00083192 Molecular Formula: C17H19N3O3S Molecular Weight: 345.42 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Melting point (°C): 151 Storage Temperature: 0-10°C

Formula: C₁₇H₁₉N₃O₃S
MW: 345.42 g/mol
Melting Pt: 151 °C
MDL Number: MFCD00083192
CAS Number: 73590-58-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3bf7ab9c2a72d680e5b774e7f0b8d57979371be9&vid=3bf7ab9c2a72d680e5b774e7f0b8d57979371be9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR