(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

O0276-1G O0276-5G
TCO0276-001GEA 84 USD
TCO0276-001G TCO0276-005G
(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid

CAS Number: 21461-84-7
MDL Number: MFCD00064516
Molecular Formula: C5H6O4
Molecular Weight: 130.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 72
Specific rotation [a]20/D: 16 deg (C=5, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-gamma-Carboxy-gamma-butyrolactone, (S)-(+)-5-Oxo-2-tetrahydrofuroic Acid

Formula: C₅H₆O₄
Melting Pt: 72 °C
MDL Number: MFCD00064516
CAS Number: 21461-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fb298c04cd51eb4eefb6e1953b9bd2187ef23a0&vid=0fb298c04cd51eb4eefb6e1953b9bd2187ef23a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR