5-Oxoazelaic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Ketoazelaic Acid, 5-Oxononanedioic Acid

O0258-1G O0258-5G
TCO0258-001GEA 293 USD
TCO0258-001G TCO0258-005G
5-Oxoazelaic acid ≥96.0% (by GC, titration analysis)
5-Oxoazelaic acid

CAS Number: 57822-06-7
MDL Number: MFCD00075542
Molecular Formula: C9H14O5
Molecular Weight: 202.21
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 113


Other Product Names / Synonyms: 5-Ketoazelaic Acid, 5-Oxononanedioic Acid

Formula: C₉H₁₄O₅
MW: 202.21 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00075542
CAS Number: 57822-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3228b946d872b49eacb6e4b94bec7d30c656a283&vid=3228b946d872b49eacb6e4b94bec7d30c656a283&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR