Octamethyltrisiloxane ≥98.0% (by GC) for synthesis

Supplier: TCI America
Danger

Synonyms: Dimethylbis(trimethylsilyloxy)silane, Poly(dimethylsiloxane), 1,1,1,3,3,5,5,5-Octamethyltrisiloxane

O0257-25ML O0257-100ML
TCO0257-025MLEA 55.94 USD
TCO0257-025ML TCO0257-100ML
Octamethyltrisiloxane ≥98.0% (by GC) for synthesis
Octamethyltrisiloxane

CAS Number: 107-51-7 MDL Number: MFCD00008264 Molecular Formula: C8H24O2Si3 Molecular Weight: 236.53 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 153 Flash Point (°C): 39 Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Dimethylbis(trimethylsilyloxy)silane

Formula: [(CH₃)₃SiO]₂Si(CH₃)₂
MW: 236.53 g/mol
Boiling Pt: 153
Melting Pt: -82
Density: 0.82 (20 °C)
Flash Pt: 39 °C
MDL Number: MFCD00008264
CAS Number: 107-51-7
EINECS: 203-497-4
UN: 1993
ADR: 3 III
Merck Index: 12,06844
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46eaaca8ad281528ad746bb60264e269a9c1ba95&vid=46eaaca8ad281528ad746bb60264e269a9c1ba95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR