2,2',3,3',5,5',6,6'-Octafluorobiphenyl ≥97.0%
Supplier: TCI America
Synonyms:
4H,4'H-Octafluorobiphenyl, 2,2',3,3',5,5',6,6'-Octafluoro-1,1'-biphenyl
O0182-1G
O0182-5G
TCO0182-001GEA
31
USD
TCO0182-001G
TCO0182-5G
2,2',3,3',5,5',6,6'-Octafluorobiphenyl ≥97.0%
2,2',3,3',5,5',6,6'-Octafluorobiphenyl
CAS Number: 3883-86-1
MDL Number: MFCD00000312
Molecular Formula: C12H2F8
Molecular Weight: 298.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 85
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4H,4'H-Octafluorobiphenyl
Formula:
C₁₂H₂F₈ MW: 298.14 g/mol Melting Pt: 85 °C |
MDL Number:
MFCD00000312 CAS Number: 3883-86-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...