Octyltrimethylammonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Trimethyl-n-octylammonium chloride, n-Octyltrimethylammonium chloride, N,N,N-Trimethyloctan-1-aminium chloride

O0178-25G O0178-100G O0178-5G
TCO0178-025GEA 120.3 USD
TCO0178-025G TCO0178-100G TCO0178-5G
Octyltrimethylammonium chloride ≥98.0% (by titrimetric analysis)
Octyltrimethylammonium chloride

CAS Number: 10108-86-8
MDL Number: MFCD00059977
Molecular Formula: C11H26ClN
Molecular Weight: 207.79
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Trimethyl-n-octylammonium Chloride

Formula: C₁₁H₂₆ClN
MW: 207.7838 g/mol
MDL Number: MFCD00059977
CAS Number: 10108-86-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ef15817b20c177d7e0750a9b710589cd94b9bca&vid=984cabbc89762fb34626d8eb6510f1dd002b6896&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR