Octaethylene glycol monododecylether (C12E8) ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3,6,9,12,15,18,21,24-Octaoxahexatriacontan-1-ol, Octaethylene glycol monododecyl ether, Octaethylene glycol monolauryl ether
O0139-5G
O0139-1G
TCO0139-005GEA
728.56
USD
TCO0139-005G
TCO0139-1G
Octaethylene glycol monododecylether (C12E8) ≥97.0% (by GC)
Octaethylene glycol monododecylether (C12E8)
CAS Number: 3055-98-9
MDL Number: MFCD00043234
Molecular Formula: C28H58O9
Molecular Weight: 538.76
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Freezing point (°C): 32
Formula:
C₂₈H₅₈O₉ MW: 538.754 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00043234 CAS Number: 3055-98-9 |
Specification Test Results
Appearance | White or Colourless to Almost white or Almost Colourless powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...