DL-Aminoglutethimide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

A1947-5G A1947-25G A1947-1G
TCA1947-5GEA 90.57 USD
TCA1947-5G TCA1947-25G TCA1947-1G
DL-Aminoglutethimide ≥98.0% (by HPLC, titration analysis)
DL-Aminoglutethimide

CAS Number: 125-84-8
MDL Number: MFCD00010122
Molecular Formula: C13H16N2O2
Molecular Weight: 232.28
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 152


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antitumor Ingredients
Other Product Names / Synonyms: 3-(4-Aminophenyl)-3-ethylpiperidine-2,6-dione

Formula: C₁₃H₁₆N₂O₂
Melting Pt: 152 °C
MDL Number: MFCD00010122
CAS Number: 125-84-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=434e0d41048b843b7b448ab5ef6633bef1b77bac&vid=434e0d41048b843b7b448ab5ef6633bef1b77bac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR