L(+)-Ornithine dihydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: (S)-2,5-Diaminovaleric acid dihydrochloride, H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic acid dihydrochloride, L-Ornithine dihydrochloride

O0089-5G O0089-1G
TCO0089-005GEA 90 USD
TCO0089-005G TCO0089-001G
L(+)-Ornithine dihydrochloride ≥98.0% (by total nitrogen basis)
L(+)-Ornithine dihydrochloride

CAS Number: 6211-16-1
MDL Number: MFCD00066571
Molecular Formula: C5H12N2O2
Molecular Weight: 205.08
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Specific rotation [a]20/D: 18 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-Orn-OH.2HCl, (S)-2,5-Diaminopentanoic Acid Dihydrochloride, (S)-2,5-Diaminovaleric Acid Dihydrochloride

Formula: C₅H₁₄Cl₂N₂O₂
MW: 205.08 g/mol
MDL Number: MFCD00066571
CAS Number: 6211-16-1
EINECS: 228-277-5

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR