Diethyl 2-oxobutanedioate ≥95.0%
Supplier: TCI America
Synonyms:
Diethyl oxalacetate, Oxalacetic acid diethyl ester
O0073-5G
O0073-25G
TCO0073-005GEA
132
USD
TCO0073-005G
TCO0073-025G
Diethyl 2-oxobutanedioate ≥95.0%
Diethyl 2-oxobutanedioate
CAS Number: 108-56-5
MDL Number: MFCD00059229
Molecular Formula: C8H12O5
Molecular Weight: 188.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 87
Specific Gravity (20/20): 1.14
Storage Temperature: <0°C
Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: Ethyl Oxalacetate, Oxalacetic Acid Diethyl Ester
Formula:
C₈H₁₂O₅ MW: 188.18 g/mol Boiling Pt: 87 °C Density: 1.14 (20 °C) |
MDL Number:
MFCD00059229 CAS Number: 108-56-5 |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...