Oleyl amine 50.0% (by GC)

Supplier: TCI America

Synonyms: cis-9-Octadecenylamine, (9Z)-9-Octadecen-1-amine, (Z)-Octadec-9-en-1-amine

O0059-500ML O0059-25ML
TCO0059-500MLEA 127.37 USD
TCO0059-500ML TCO0059-025ML
Oleyl amine 50.0% (by GC)
Oleyl amine

CAS Number: 112-90-3
MDL Number: MFCD00066507
Molecular Formula: C18H37N
Molecular Weight: 267.50
Purity/Analysis Method: >50.0% (GC)
Form: Clear Liquid
Flash Point (°C): 158
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: cis-9-Octadecenylamine

Formula: C₁₈H₃₇N
MW: 267.5 g/mol
Boiling Pt: 364 °C (1013 hPa)
Melting Pt: 21 °C
Density: 0.83 (20 °C)
Flash Pt: 158 °C
MDL Number: MFCD00066507
CAS Number: 112-90-3
EINECS: 204-015-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aea92ea7e4ab5fcbff6381e6d8c20b118d771162&vid=aea92ea7e4ab5fcbff6381e6d8c20b118d771162&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >50.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR