Sodium octanoate ≥99.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Octanoic acid sodium acid, Sodium caprylate, Caprylic acid sodium acid, Sodium n-Octanoate
O0034-250G
O0034-25G
TCO0034-250GEA
149.33
USD
TCO0034-250G
TCO0034-025G
Sodium octanoate ≥99.0% (by titrimetric analysis)
Sodium octanoate
CAS Number: 1984-06-1
MDL Number: MFCD00058511
Molecular Formula: C8H16O2
Molecular Weight: 166.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: n-Octanoic Acid Sodium Salt
Formula:
C₈H₁₅NaO₂ MW: 166.2 g/mol Melting Pt: 225…355 °C Density: Min. 0.9949 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00058511 CAS Number: 1984-06-1 EINECS: 217-850-5 |
Specification Test Results
Purity/Analysis Method | >99.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...