Isobutyl n-octanoate ≥99.0%

Supplier: TCI America

Synonyms: n-Octanoic Acid Isobutyl Ester

O0032-25ML O0032-500ML
TCO0032-025MLEA 57 USD
TCO0032-025ML TCO0032-500ML
Isobutyl n-octanoate ≥99.0%
Isobutyl n-octanoate

CAS Number: 5461-06-3
MDL Number: MFCD00059444
Molecular Formula: C12H24O2
Molecular Weight: 200.32
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Flash Point (°C): 108
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: n-Octanoic Acid Isobutyl Ester

Formula: C₁₂H₂₄O₂
MW: 200.32 g/mol
Density: 0.86 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00059444
CAS Number: 5461-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f5529d4f9f4c0fe5a0ef6289f3c20a3e51764ed&vid=4f5529d4f9f4c0fe5a0ef6289f3c20a3e51764ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR