Octadecylamine acetate 60.0%

Supplier: TCI America

Synonyms: Stearylamine Acetate, Aminooctadecane Acetate

O0015-25G O0015-500G
TCO0015-025GEA 17 USD
TCO0015-025G TCO0015-500G
Octadecylamine acetate 60.0%
Octadecylamine acetate

CAS Number: 2190-04-7
MDL Number: MFCD00060198
Molecular Formula: C18H39N
Molecular Weight: 329.57
Purity/Analysis Method: >60.0% (GC)
Form: Crystal
Melting point (°C): 75


Other Product Names / Synonyms: Aminooctadecane Acetate, Stearylamine Acetate

Formula: C₁₈H₃₉N
MW: 329.57 g/mol
Melting Pt: 75 °C
MDL Number: MFCD00060198
CAS Number: 2190-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ee27c9df721ee9a935e08c55ea12da66b6658cf&vid=7ee27c9df721ee9a935e08c55ea12da66b6658cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >60.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR