4-Amino-5-chloro-2-ethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Amino-5-chloro-2-ethoxybenzoic Acid

A1941-25G A1941-5G
TCA1941-25GEA 77 USD
TCA1941-25G TCA1941-5G
4-Amino-5-chloro-2-ethoxybenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Amino-5-chloro-2-ethoxybenzoic acid

CAS Number: 108282-38-8
MDL Number: MFCD03840527
Molecular Formula: C9H10ClNO3
Molecular Weight: 215.63
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 169

Formula: C₉H₁₀ClNO₃
MW: 215.63 g/mol
Melting Pt: 169 °C
MDL Number: MFCD03840527
CAS Number: 108282-38-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09b70f9ecb0946c943c78334a980d6ffa5017e49&vid=09b70f9ecb0946c943c78334a980d6ffa5017e49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR