5-(4-Nitrophenyl)-2-furaldehyde ≥98.0%

Supplier: TCI America

Synonyms: 5-(4-Nitrophenyl)furan-2-carbaldehyde, Dantrolene Related Compound C, 5-(4-Nitrophenyl)furfural

N0820-25G N0820-5G
TCN0820-25GEA 174 USD
TCN0820-25G TCN0820-5G
5-(4-Nitrophenyl)-2-furaldehyde ≥98.0%
5-(4-Nitrophenyl)-2-furaldehyde

CAS Number: 7147-77-5
MDL Number: MFCD00124191
Molecular Formula: C11H7NO4
Molecular Weight: 217.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 207


Other Product Names / Synonyms: 5-(4-Nitrophenyl)furfural

Formula: C₁₁H₇NO₄
Melting Pt: 207 °C
MDL Number: MFCD00124191
CAS Number: 7147-77-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cf4ea2022d0e31e4f98f9e453926b9beffcf82a&vid=8cf4ea2022d0e31e4f98f9e453926b9beffcf82a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR