3-Noradamantanamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

N0778-1G N0778-5G
TCN0778-1GEA 106 USD
TCN0778-1G TCN0778-5G
3-Noradamantanamine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
3-Noradamantanamine hydrochloride

CAS Number: 86128-83-8
MDL Number: MFCD00213499
Molecular Formula: C9H15N
Molecular Weight: 173.68
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 350


Other Product Names / Synonyms: 3-Aminonoradamantane Hydrochloride

Formula: C₉H₁₅N
Melting Pt: 350 °C
MDL Number: MFCD00213499
CAS Number: 86128-83-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=415c3bc1a6cba698e0f869449795119176a0f599&vid=415c3bc1a6cba698e0f869449795119176a0f599&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR