2-Nitrophenyl phenyl sulfide ≥98.0%

Supplier: TCI America

N0744-5G N0744-25G
TCN0744-5GEA 116.98 USD
TCN0744-5G TCN0744-25G
2-Nitrophenyl phenyl sulfide ≥98.0%
2-Nitrophenyl phenyl sulphide

CAS Number: 4171-83-9
Molecular Formula: C12H9NO2S
Molecular Weight: 231.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 82


Other Product Names / Synonyms: 2-Nitrodiphenyl Sulfide, 1-Nitro-2-(phenylthio)benzene

Formula: C₁₂H₉NO₂S
Melting Pt: 82 °C
CAS Number: 4171-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a2ca484fa6f55229232b54cbebd9d543a5a7efc&vid=2a2ca484fa6f55229232b54cbebd9d543a5a7efc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR