(R)-(-)-Ethyl-3-piperidinecarboxylate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl (R)-(-)-3-Piperidinecarboxylate, (R)-Ethyl piperidine-3-carboxylate

N0655-25G N0655-5G
TCN0655-25GEA 390 USD
TCN0655-25G TCN0655-5G
(R)-(-)-Ethyl-3-piperidinecarboxylate ≥98.0%
(R)-(-)-Ethyl 3-piperidinecarboxylate

CAS Number: 25137-01-3
MDL Number: MFCD03093637
Molecular Formula: C8H15NO2
Molecular Weight: 157.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 90
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: -1.5 deg (neat)
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl (R)-(-)-Nipecotate, (R)-(-)-Nipecotic Acid Ethyl Ester, (R)-(-)-3-Piperidinecarboxylic Acid Ethyl Ester

Formula: C₈H₁₅NO₂
MW: 157.2102 g/mol
Boiling Pt: 110 °C
Density: 1.02 (20 °C)
Flash Pt: 90 °C
MDL Number: MFCD03093637
CAS Number: 25137-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd0407e410fba00dbe218c179d72e42841fa4674&vid=bd0407e410fba00dbe218c179d72e42841fa4674&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR