4-(4-Nitrophenylazo)phenol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4-Hydroxy-4'-nitroazobenzene
N0653-1G
N0653-5G
TCN0653-1GEA
285
USD
TCN0653-1G
TCN0653-005G
4-(4-Nitrophenylazo)phenol ≥97.0% (by GC, titration analysis)
4-(4-Nitrophenylazo)phenol
CAS Number: 1435-60-5
MDL Number: MFCD00161165
Molecular Formula: C12H9N3O3
Molecular Weight: 243.22
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: 4-Hydroxy-4'-nitroazobenzene
Formula:
C₁₂H₉N₃O₃ MW: 243.22 g/mol |
MDL Number:
MFCD00161165 CAS Number: 1435-60-5 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...