4-Nitrophenylethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-(4-Nitrophenyl)ethylamine hydrochloride

N0648-5G N0648-25G
TCN0648-005GEA 22 USD
TCN0648-005G TCN0648-25G
4-Nitrophenylethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitrophenylethylamine hydrochloride

CAS Number: 29968-78-3
MDL Number: MFCD00012900
Molecular Formula: C8H10N2O2
Molecular Weight: 202.64
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 214


Other Product Names / Synonyms: 4-Nitrophenethylamine Hydrochloride

Formula: C₈H₁₁ClN₂O₂
MW: 202.64 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00012900
CAS Number: 29968-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=28dfb48fb9b96f00507edfa6b1cf1adbbc1fce9d&vid=28dfb48fb9b96f00507edfa6b1cf1adbbc1fce9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR