2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate

Supplier: TCI America

Synonyms: TNTU, 2-(1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate

N0634-5G N0634-25G
TCN0634-005GEA 41 USD
TCN0634-005G TCN0634-025G
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate

[Coupling Reagent for Peptide]
CAS Number: 125700-73-4
MDL Number: MFCD00167753
Molecular Formula: C14H20BF4N3O3
Molecular Weight: 365.14
Form: Crystal
Color: White


Other Product Names / Synonyms: TNTU

Formula: C₁₄H₂₀BF₄N₃O₃
MDL Number: MFCD00167753
CAS Number: 125700-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=589099041dcb9bf3b23b7c46f7837ee92cf79911&vid=589099041dcb9bf3b23b7c46f7837ee92cf79911&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR