1,2-Naphthalic anhydride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,2-Naphthalenedicarboxylic Anhydride

N0583-25G N0583-5G
TCN0583-25GEA 1815 USD
TCN0583-25G TCN0583-005G
1,2-Naphthalic anhydride ≥98.0% (by GC, titration analysis)
1,2-Naphthalic anhydride

CAS Number: 5343-99-7
MDL Number: MFCD00191561
Molecular Formula: C12H6O3
Molecular Weight: 198.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 170


Other Product Names / Synonyms: 1,2-Naphthalenedicarboxylic Anhydride

Formula: C₁₂H₆O₃
MW: 198.18 g/mol
Melting Pt: 170 °C
MDL Number: MFCD00191561
CAS Number: 5343-99-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cb122b9f466e87301ad331a073debbef07728a0&vid=0cb122b9f466e87301ad331a073debbef07728a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR