2-(1-Naphthyl)ethanesulfonyl chloride ≥97.0%
Supplier: TCI America
Synonyms:
2-(1-Naphthyl)ethylsulphonyl Chloride
N0576-5G
N0576-1G
TCN0576-005GEA
192.5
USD
TCN0576-005G
TCN0576-001G
2-(1-Naphthyl)ethanesulfonyl chloride ≥97.0%
2-(1-Naphthyl)ethanesulfonyl chloride
CAS Number: 104296-63-1
MDL Number: MFCD00191560
Molecular Formula: C12H11ClO2S
Molecular Weight: 254.73
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 50
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: 2-(1-Naphthyl)ethylsulfonyl Chloride
Formula:
C₁₂H₁₁ClO₂S Melting Pt: 50 °C |
MDL Number:
MFCD00191560 CAS Number: 104296-63-1 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...