4'-Nitro-3'-(trifluoromethyl)acetanilide ≥98.0%
Supplier: TCI America
Synonyms:
N-(4-Nitro-3-(trifluoromethyl)phenyl)acetamide
N0548-25G
N0548-5G
TCN0548-025GEA
213.4
USD
TCN0548-025G
TCN0548-005G
4'-Nitro-3'-(trifluoromethyl)acetanilide ≥98.0%
4'-Nitro-3'-(trifluoromethyl)acetanilide
CAS Number: 393-12-4
MDL Number: MFCD00017994
Molecular Formula: C9H7F3N2O3
Molecular Weight: 248.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 123
Formula:
C₉H₇F₃N₂O₃ MW: 248.1600 g/mol Melting Pt: 123 °C |
MDL Number:
MFCD00017994 CAS Number: 393-12-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...