Nifedipine ≥98.0%

Supplier: TCI America

Synonyms: Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate, Adalat, 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester

N0528-10G N0528-25G
TCN0528-010GEA 80.5 USD
TCN0528-010G TCN0528-25G
Nifedipine ≥98.0%
Nifedipine

CAS Number: 21829-25-4
MDL Number: MFCD00057326
Molecular Formula: C17H18N2O6
Molecular Weight: 346.34
Purity/Analysis Method: >98.0% (HPLC,E)
Form: Crystal
Melting point (°C): 174


Product Categories: Materials Chemistry, Polymerization Initiators, Bioscience, Antihypertensive Ingredients, Voltage-dependent Calcium Channel (VDCC) Antagonists
Other Product Names / Synonyms: 1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester

Formula: C₁₇H₁₈N₂O₆
MW: 346.34 g/mol
Melting Pt: 174 °C
MDL Number: MFCD00057326
CAS Number: 21829-25-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906c11cc12962e71176c5bcb90727134af94d593&vid=906c11cc12962e71176c5bcb90727134af94d593&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR