Nicotinic acid N-oxide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Oxiniacic Acid, 3-Carboxypyridine 1-oxide

N0497-25G N0497-500G
TCN0497-025GEA 46 USD
TCN0497-025G TCN0497-500G
Nicotinic acid N-oxide ≥98.0% (by titrimetric analysis)
Nicotinic acid N-oxide

CAS Number: 2398-81-4
MDL Number: MFCD00006201
Molecular Formula: C6H5NO3
Molecular Weight: 139.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 255


Other Product Names / Synonyms: Oxiniacic Acid

Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00006201
CAS Number: 2398-81-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbe7ff86d77feafb5966e0f41274f1c7a31cd64a&vid=cbe7ff86d77feafb5966e0f41274f1c7a31cd64a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR