2-Nonene (cis- and trans- mixture) cis- and trans-mixture ≥94.0% (by GC)

Supplier: TCI America
Danger

N0476-5ML N0476-1ML
TCN0476-005MLEA 883 USD
TCN0476-005ML TCN0476-1ML
2-Nonene (cis- and trans- mixture) cis- and trans-mixture ≥94.0% (by GC)
2-Nonene (cis- and trans- mixture)

CAS Number: 2216-38-8
MDL Number: MFCD00009549
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: >94.0% (GC)
Form: Clear Liquid
Flash Point (°C): 32
Specific Gravity (20/20): 0.74
Storage Temperature: 0-10°C

Formula: C₉H₁₈
Density: 0.74 (20 °C)
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009549
CAS Number: 2216-38-8
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 94.0% (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR