1,6-Naphthyridine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 253-72-5
MDL Number: MFCD00059750
Molecular Formula: C8H6N2
Molecular Weight: 130.15
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Freezing point (°C): 35
Other Product Names / Synonyms: 1,6-Diazanaphthalene, Pyrido[4,3-b]pyridine
Formula:
C₈H₆N₂ |
MDL Number:
MFCD00059750 CAS Number: 253-72-5 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...