6-Acrylamidohexanoic acid 98,0

Supplier: TCI America

Synonyms: 6-Acrylamidohexanoic Acid

A1896-1G A1896-5G A1896-25G
TCA1896-1GEA 48 USD
TCA1896-1G TCA1896-5G TCA1896-25G
6-Acrylamidohexanoic acid 98,0
6-Acrylamidohexanoic acid

CAS Number: 20766-85-2
Molecular Formula: C9H15NO3
Molecular Weight: 185.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 89

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Melting Pt: 89 °C
MDL Number: MFCD06804452
CAS Number: 20766-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f70fad9035e5733a5a52b55efb6028b88929ddea&vid=f70fad9035e5733a5a52b55efb6028b88929ddea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR