Norethisterone acetate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
CAS Number: 51-98-9
MDL Number: MFCD04039850
Molecular Formula: C22H28O3
Molecular Weight: 340.46
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -34 deg (C=1, Dioxane)
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Bioscience, Steroids, Other Steroids
Other Product Names / Synonyms: 17alpha-Ethynyl-19-nortestosterone Acetate, 19-Nor-17alpha-ethynyltestosterone Acetate
Formula:
C₂₂H₂₈O₃ |
MDL Number:
MFCD04039850 CAS Number: 51-98-9 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...