Neostigmine methyl sulfate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
(3-Dimethylcarbamoyloxyphenyl)trimethylammonium Methyl Sulphate, 3-((Dimethylcarbamoyl)oxy)-N,N,N-trimethylbenzenaminium methyl sulphate, (3-Hydroxyphenyl)trimethylammonium Methyl sulphate Dimethylcarbamate
CAS Number: 51-60-5
MDL Number: MFCD00011796
Molecular Formula: C13H22N2O6S
Molecular Weight: 334.39
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 146
Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Methyl Sulfate, (3-Hydroxyphenyl)trimethylammonium Methyl Sulfate Dimethylcarbamate
Formula:
C₁₃H₂₂N₂O₆S MW: 334.39 g/mol Melting Pt: 146 °C |
MDL Number:
MFCD00011796 CAS Number: 51-60-5 |
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...