Sodium naphthenate
Supplier: TCI America
Synonyms:
Naphthenic Acid Sodium Salt
N0397-25G
N0397-500G
TCN0397-025GEA
41.41
USD
TCN0397-025G
TCN0397-500G
Sodium naphthenate
Sodium naphthenate
CAS Number: 61790-13-4
MDL Number: MFCD00147698
Form: Crystal
Color: Very Pale Yellow
Other Product Names / Synonyms: Naphthenic Acid Sodium Salt
MDL Number:
MFCD00147698 CAS Number: 61790-13-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...