DL-Norphenylephrine hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
3-(2-Amino-1-hydroxyethyl)phenol Hydrochloride
N0389-5G
N0389-25G
TCN0389-5GEA
71
USD
TCN0389-5G
TCN0389-025G
DL-Norphenylephrine hydrochloride ≥98.0%
DL-Norphenylephrine hydrochloride
CAS Number: 4779-94-6
MDL Number: MFCD00012879
Molecular Formula: C8H11NO2
Molecular Weight: 189.64
Purity/Analysis Method:
Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: 3-(2-Amino-1-hydroxyethyl)phenol Hydrochloride
Formula:
C₈H₁₁NO₂ Melting Pt: 157 °C |
MDL Number:
MFCD00012879 CAS Number: 4779-94-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...