Neostigmine bromide ≥99.0%

Supplier: TCI America

Synonyms: 3-((Dimethylcarbamoyl)oxy)-N,N,N-trimethylbenzenaminium bromide, (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Bromide, (3-Hydroxyphenyl)trimethylammonium Bromide Dimethyl Carbamate

N0358-5G N0358-25G
TCN0358-005GEA 94.2 USD
TCN0358-005G TCN0358-025G
Neostigmine bromide ≥99.0%
Neostigmine bromide

CAS Number: 114-80-7
MDL Number: MFCD00011795
Molecular Formula: C12H19BrN2O2
Molecular Weight: 303.20
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: (3-Dimethylcarbamoyloxyphenyl)trimethylammonium Bromide, (3-Hydroxyphenyl)trimethylammonium Bromide Dimethyl Carbamate

Formula: C₁₂H₁₉BrN₂O₂
MW: 303.20 g/mol
MDL Number: MFCD00011795
CAS Number: 114-80-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6019ef46c8ef97859259f47646d1eac127cde617&vid=6019ef46c8ef97859259f47646d1eac127cde617&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR