Naphthenic acid

Supplier: TCI America

Synonyms: Naphthenic acids

N0343-25G N0343-500G
TCN0343-025GEA 35.6 USD
TCN0343-025G TCN0343-500G
Naphthenic acid
Naphthenic acid

CAS Number: 1338-24-5
MDL Number: MFCD00147696
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 165
Specific Gravity (20/20): 0.96

Density: 0.96 (20 °C)
Flash Pt: 165 °C
MDL Number: MFCD00147696
CAS Number: 1338-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR