3,3',3''-Nitrilotris(propionamide) ≥98.0% (by titrimetric analysis)

Supplier: TCI America

N0342-25G N0342-500G
TCN0342-025GEA 71.5 USD
TCN0342-025G TCN0342-500G
3,3',3''-Nitrilotris(propionamide) ≥98.0% (by titrimetric analysis)
3,3',3''-Nitrilotris(propionamide)

CAS Number: 2664-61-1
MDL Number: MFCD00059194
Molecular Formula: C9H18N4O3
Molecular Weight: 230.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 183

Formula: C₉H₁₈N₄O₃
MW: 230.27 g/mol
Melting Pt: 183 °C
MDL Number: MFCD00059194
CAS Number: 2664-61-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9360e6a4a47fb28bfa92f49ddf1081b9cd77218b&vid=9360e6a4a47fb28bfa92f49ddf1081b9cd77218b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR