Manganese naphthenate (∼6% Mn)
Supplier: TCI America
Synonyms:
Naphthenic Acid Manganese Salt
N0340-25G
N0340-500G
TCN0340-025GEA
35.82
USD
TCN0340-025G
TCN0340-500G
Manganese naphthenate (∼6% Mn)
Manganese naphthenate
CAS Number: 1336-93-2
MDL Number: MFCD00147533
Form: Liquid
Color: Black
Flash Point (°C): 55
Other Product Names / Synonyms: Naphthenic Acid Manganese Salt
Formula:
₂(C₁₁H₇O₂).Mn MW: 397.29 g/mol Flash Pt: 55 °C |
MDL Number:
MFCD00147533 CAS Number: 1336-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...