4-(4-Nitrophenylazo)orcinol ≥80.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-Nitrobenzene-azo-orcinol, 4-(4-Nitrophenylazo)-5-methylresorcinol

N0324-5G N0324-1G
TCN0324-5GEA 754 USD
TCN0324-5G TCN0324-001G
4-(4-Nitrophenylazo)orcinol ≥80.0% (by HPLC)
4-(4-Nitrophenylazo)orcinol

CAS Number: 607-96-5
MDL Number: MFCD00014710
Molecular Formula: C13H11N3O4
Molecular Weight: 273.25
Purity/Analysis Method: >80.0% (HPLC)
Form: Crystal


Other Product Names / Synonyms: 4-Nitrobenzene-azo-orcinol, 4-(4-Nitrophenylazo)-5-methylresorcinol

Formula: C₁₃H₁₁N₃O₄
MW: 273.25 g/mol
MDL Number: MFCD00014710
CAS Number: 607-96-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8830838de06723b4ac2fe09ebe27f0b8488e1967&vid=8830838de06723b4ac2fe09ebe27f0b8488e1967&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >80.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR