4-Nitrotoluene-2-sulfonic acid hydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
p-Nitrotoluene-o-sulphonic acid
N0277-500G
N0277-25G
TCN0277-500GEA
316
USD
TCN0277-500G
TCN0277-025G
4-Nitrotoluene-2-sulfonic acid hydrate ≥95.0% (by titrimetric analysis)
4-Nitrotoluene-2-sulfonic acid hydrate
CAS Number: 121-03-9
MDL Number: MFCD00024914
Molecular Formula: C7H7NO5S
Molecular Weight: 217.20
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 135
Formula:
C₇H₇NO₅S MW: 217.20 g/mol Melting Pt: 135 °C |
MDL Number:
MFCD00024914 CAS Number: 121-03-9 |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...