Sodium-3-nitrosalicylate ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Nitrosalicylic Acid Sodium Salt

N0258-5G N0258-25G
TCN0258-005GEA 59 USD
TCN0258-005G TCN0258-025G
Sodium-3-nitrosalicylate ≥99.0% (by titrimetric analysis)
Sodium-3-nitrosalicylate

CAS Number: 164915-53-1
MDL Number: MFCD00070529
Molecular Formula: C7H5NO5
Molecular Weight: 205.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal


Other Product Names / Synonyms: 3-Nitrosalicylic Acid Sodium Salt

Formula: C₇H₅NO₅
MW: 205.10 g/mol
MDL Number: MFCD00070529
CAS Number: 164915-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=095e2fe7a335d43982711c836b9e5fb3d9a101d3&vid=095e2fe7a335d43982711c836b9e5fb3d9a101d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR