3-Nitrosalicylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Nitrosalicylic Acid, 2-Hydroxy-3-nitrobenzoic acid

N0251-25G N0251-5G
TCN0251-025GEA 142 USD
TCN0251-025G TCN0251-5G
3-Nitrosalicylic acid ≥98.0% (by GC, titration analysis)
3-Nitrosalicylic acid

CAS Number: 85-38-1
MDL Number: MFCD00024240
Molecular Formula: C7H5NO5
Molecular Weight: 183.12
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 149

Formula: C₇H₅NO₅
MW: 183.1183 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00024240
CAS Number: 85-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cded5a1673a4780e230ff817fc9776c6ff6b5132&vid=cded5a1673a4780e230ff817fc9776c6ff6b5132&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR