(3-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
N0224-5G
N0224-25G
TCN0224-005GEA
65.9
USD
TCN0224-005G
TCN0224-025G
(3-Nitrophenoxy)acetic acid ≥98.0% (by GC, titration analysis)
(3-Nitrophenoxy)acetic acid
CAS Number: 1878-88-2
MDL Number: MFCD00059826
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >98.0% (T,GC)
Form: Crystal
Melting point (°C): 153
Formula:
(C₆H₄(NO₂))OCH₂COOH Melting Pt: 153 °C |
MDL Number:
MFCD00059826 CAS Number: 1878-88-2 EINECS: 217-528-4 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T,GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...