2-Nitrophenol sodium salt ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Sodium 2-Nitrophenol
N0222-25G
N0222-500G
TCN0222-025GEA
32
USD
TCN0222-025G
TCN0222-500G
2-Nitrophenol sodium salt ≥99.0% (by HPLC, titration analysis)
2-Nitrophenol sodium salt
CAS Number: 824-39-5
MDL Number: MFCD00065178
Molecular Formula: C6H5NO3
Molecular Weight: 161.09
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Product Categories: Analytical Science, Indicators (pH)
Other Product Names / Synonyms: Sodium 2-Nitrophenol
Formula:
C₆H₅NO₃ MW: 161.09 g/mol |
MDL Number:
MFCD00065178 CAS Number: 824-39-5 |
|
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC,T) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...