n-Nonacosane ≥98.0%

Supplier: TCI America

Synonyms: Nonacosane

N0167-1G N0167-100MG
TCN0167-001GEA 381.22 USD
TCN0167-001G TCN0167-100MG
n-Nonacosane ≥98.0%
n-Nonacosane

CAS Number: 630-03-5
MDL Number: MFCD00009356
Molecular Formula: C29H60
Molecular Weight: 408.80
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 64

Formula: H₃C(CH₂)₂₇CH₃
MW: 408.8 g/mol
Boiling Pt: 440.8 °C (1013 hPa)
Melting Pt: 64 °C
Density: 0.8083 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009356
CAS Number: 630-03-5
EINECS: 211-126-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cf7c23d33f0c92f187e004a0bdfc38f3872ccc7&vid=19ef4e957a91139ec41eb254e9f241e7b55e5019&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR