N-Nicotinoylglycine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-(Nicotinamido)acetic acid, Nicotinuric Acid
N0092-25G
N0092-5G
TCN0092-025GEA
401
USD
TCN0092-025G
TCN0092-005G
N-Nicotinoylglycine ≥98.0% (by titrimetric analysis)
N-Nicotinoylglycine
CAS Number: 583-08-4
MDL Number: MFCD00023578
Molecular Formula: C8H8N2O3
Molecular Weight: 180.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nicotinuric Acid
Formula:
C₈H₈N₂O₃ MW: 180.16 g/mol |
MDL Number:
MFCD00023578 CAS Number: 583-08-4 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...