(S)-(-)-Nicotine ≥95.0% (by GC, titration analysis)

Supplier: TCI America
Danger

N0079-25ML N0079-500ML N0079-100ML
TCN0079-025MLEA 67.13 USD
TCN0079-025ML TCN0079-500ML TCN0079-100ML
(S)-(-)-Nicotine ≥95.0% (by GC, titration analysis)

CAS Number: 54-11-5
MDL Number: MFCD00006369
Molecular Formula: C10H14N2
Molecular Weight: 162.24
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 91
Melting point (°C): -80
Flash Point (°C): 95
Specific Gravity (20/20): 1.02
Specific rotation [a]20/D: -167 deg (C=10, Acetone)


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Alkaloids (Phytochemicals), Pyridine Alkaloids
Other Product Names / Synonyms: (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine

Formula: C₁₀H₁₄N₂
Boiling Pt: 91 °C
Melting Pt: –80 °C
Density: 1.02 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00006369
CAS Number: 54-11-5
EINECS: 200-193-3
UN: 1654
ADR: 6.1,II
Merck Index: 12,06611
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=24ebd94cc59544cf16e970ee78f7a87883491cb9&vid=24ebd94cc59544cf16e970ee78f7a87883491cb9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR